Order Entry
Denmark
ContactUsLinkComponent
 
Phosphate free detergent no foam, RBS 50 PF
  CHEPRBS/50/PF/5L
 :  CHEMICAL
undefined
Phosphate free detergent no foam, RBS 50 PF
  CHEPRBS/50/PF/5L
 :  CHEMICAL

 

  • Description:
    Phosphate free detergent no foam, RBS 50 PF
  • Pk:
    5 L
  • pH range:
    11.3 (1%)

 

 

Efficient cleaning of glassware and laboratory equipment in automatic washing machines - labwashers.

  • High detergency power
  • Phosphate-free
  • Contains active chlorine
  • Low use concentration
  • Easy rinse

RBS 50 PF is recommended for cleaning glassware and laboratory equipment in automatic washing machine - labwashers. It efficiently removes organic and biological residues, distilation residues etc. Particularly adapted for use in water analysis and environmental laboratories.

 : Available in box of 4 × 5 litres. Please contact Avantor for details.
 : Not recommended for use on white metals.