Dehydrated culture media, superbroth, Molecular Biology Certified™, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Superbroth is a rich medium with high levels of tryptone and yeast extract used for obtaining high yields of E. coli cells and lambda bacteriophages.
- Available as capsules or powder
- Molecular biology certified
Due to the high levels of peptides, amino acids, vitamins and cofactors high yields of lambda phage and phagemids in E. coli are obtained.
Frequently Bought Together