Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffers, concentrated solutions, Fluka™
Buffers, concentrated solutions, Fluka™
Buffers, concentrated solutions, Fluka™
Certificates
Buffers, concentrated solutions, Fluka™
16 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • pH value
    pH accuracy
    Contents
    Final volume
    Pack type
    Pk
    Availability
    Price
  • 1,00
    ±0.02
    hydrochloric acid/potassium chloride
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 413,00 kr
    413,00 kr
    1 * 1 Items
       413,00 kr
  • 2,00
    ±0.02
    citric acid/hydrochloric acid/sodium chloride
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 473,00 kr
    473,00 kr
    1 * 1 Items
       473,00 kr
  • 3,00
    ±0.02
    citric acid/sodium hydroxide/sodium chloride solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 384,00 kr
    384,00 kr
    1 * 1 Items
       384,00 kr
  • 4,00
    ±0.02
    citric acid/sodium hydroxide/sodium chloride solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 220,00 kr
    220,00 kr
    1 * 1 Items
       220,00 kr
  • 4,00
    ±0.02
    citric acid/sodium hydroxide/sodium chloride solution
    500 ml
    Ampoule
    6
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 1.250,00 kr
    1.250,00 kr
    1 * 6 Items
       1.250,00 kr
  • 5,00
    ±0.02
    citric acid/sodium hydroxide solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 546,00 kr
    546,00 kr
    1 * 1 Items
       546,00 kr
  • 7,00
    ±0.02
    potassium dihydrogen phosphate/disodium hydrogen phosphate
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 251,00 kr
    251,00 kr
    1 * 1 Items
       251,00 kr
  • 7,00
    ±0.02
    potassium dihydrogen phosphate/disodium hydrogen phosphate
    500 ml
    Ampoule
    6
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 1.420,00 kr
    1.420,00 kr
    1 * 6 Items
       1.420,00 kr
  • 8,00
    ±0.02
    sodium tetraborate/hydrochloric acid
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 635,00 kr
    635,00 kr
    1 * 1 Items
       635,00 kr
  • 9,00
    ±0.02
    sodium tetraborate/hydrochloric acid
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 221,00 kr
    221,00 kr
    1 * 1 Items
       221,00 kr
  • 9,00
    ±0.02
    sodium tetraborate/hydrochloric acid
    500 ml
    Ampoule
    6
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 1.330,00 kr
    1.330,00 kr
    1 * 6 Items
       1.330,00 kr
  • 10,00
    ±0.05
    sodium tetraborate/sodium hydroxide solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
  • 10,00
    ±0.05
    sodium tetraborate/sodium hydroxide solution
    500 ml
    Ampoule
    6
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 1.830,00 kr
    1.830,00 kr
    1 * 6 Items
       1.830,00 kr
  • 11,00
    ±0.05
    boric acid/sodium hydroxide solution/potassium chloride
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 639,00 kr
    639,00 kr
    1 * 1 Items
       639,00 kr
  • 12,00
    ±0.05
    di-sodium hydrogen phosphate/sodium hydroxide solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 542,00 kr
    542,00 kr
    1 * 1 Items
       542,00 kr
  • 13,00
    ±0.05
    glycine/sodium hydroxide/sodium chloride solution
    500 ml
    Ampoule
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 278,00 kr
    278,00 kr
    1 * 1 Items
       278,00 kr