Electrolyte solutions, organic
Supplier: SI Analytics
Jump to:
About this item
Electrolyte solutions for measurements in organic solutions for reference electrodes and as electrolyte bridges.
2 Options Available Below
Product Details & Documents
Electrolyte solutions for measurements in organic solutions for reference electrodes and as electrolyte bridges.
Documents
SI Analytics Process Equipment Catalog VWR Process Equipment Catalog.pdf
Product Literature
Product Family Options
Product Information
- PkAvailabilityPrice
- SCOI285138308Electrolyte solution, L 5034, LiCl saturated in ethanol, 250 ml in DURAN® glass bottleElectrolyte solution, L 5034, LiCl saturated in ethanol, 250 ml in DURAN® glass bottle250 mLLog in for availability1 * 250 mL974,00 kr
- SCOI285138324Electrolyte solution, L 5014, LiCl saturated in glacial acetic acid, 250 ml in DURAN® glass bottleElectrolyte solution, L 5014, LiCl saturated in glacial acetic acid, 250 ml in DURAN® glass bottle250 mLLog in for availability1 * 250 mL1.290,00 kr