Anti-DBF4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-7895R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DBF4 Zinc Finger
- Antigen symbol:DBF4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10926
- Antigen synonyms:DBF4A|CHIF|ASK|ZDBF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Regulatory subunit for CDC7 which activates its kinase activity thereby playing a central role in DNA replication and cell proliferation. Required for progression of S phase. The complex CDC7-DBF4A selectively phosphorylates MCM2 subunit at 'Ser-40' and 'Ser-53' and then is involved in regulating the initiation of DNA replication during cell cycle.Tissue specificity: Highly expressed in testis and thymus. Expressed also in most cancer cells lines.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DBF4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat