Anti-A4GALT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-7588R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Alpha 1,4-Galactosyltransferase
- Antigen symbol:A4GALT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:53947
- Antigen synonyms:A4GALT1|P(k)|P1|P1PK|Gb3S|A4GALT|PK|A14GALT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:290-340/353
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat