Order Entry
Denmark
ContactUsLinkComponent
Anti-LIMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-LIMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-7336R-A647
Supplier:  Bioss
Anti-LIMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-7336R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    LIM Domains Containing 1
  • Antigen symbol:
    LIMD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8994
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing 3 LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is down-regulated in lung cancer. Limd1 may regulate osteoclast development under stressful conditions via its interactions with TRAF6 and p62.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIMD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat