Order Entry
Denmark
ContactUsLinkComponent
Anti-KCNQ1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-KCNQ1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-6760R-A350
 :  Bioss
Anti-KCNQ1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-6760R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Potassium Voltage-gated Channel KQT-like Subfamily Member 1
  • Antigen symbol:
    KCNQ1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3784
  • Antigen synonyms:
    Kv7.1|JLNS1|FLJ26167|KCNQ1|potassium voltage-gated channel|Jervell and Lange-Nielsen syndrome 1|WRS|ATFB1|long (electrocardiographic) QT syndrome|SQT2|Ward-Romano syndrome 1|LQT1|LQT|KQT-like subfamily|Kv1.9|member 1|KCNA8|slow delayed rectifier channel subunit|KVLQT1|kidney and cardiac voltage dependend K+ channel|RWS|KCNA9
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    500-550/676
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Probably important in cardiac repolarization. Associates with KCNE1 (MinK) to form the I(Ks) cardiac potassium current. Elicits a rapidly activating, potassium-selective outward current. Muscarinic agonist oxotremorine-M strongly suppresses KCNQ1/KCNE1 current in CHO cells in which cloned KCNQ1/KCNE1 channels were coexpressed with M1 muscarinic receptors. May associate also with KCNE3 (MiRP2) to form the potassium channel that is important for cyclic AMP-stimulated intestinal secretion of chloride ions, which is reduced in cystic fibrosis and pathologically stimulated in cholera and other forms of secretory diarrhea.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KCNQ1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog