- Antibody type:Primary
- Antigen name:Lectin LCA
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Lens culinaris agglutinin is composed of four subunits - two of about 17 kDa and two of 8 kDa. LCA recognizes sequences containing α-linked mannose residues but recognizes additional sugars as part of the receptor structure, giving it a narrower specificity than Con A. An α-linked fucose residue attached to the N-acetylchitobiose portion of the core oligosaccharide markedly enhances affinity. LCA has been found to be one of the most effective agents in preventing skin allograft rejection in model systems. LCA has been used to purify numerous glycoproteins, including immunoglobulins, histocompatibility antigens, and α2-macroglobulin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Lectin LCA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species