Anti-ILK Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-5443R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Integrin-Linked Kinase
- Antigen symbol:ILK
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3611
- Antigen synonyms:ILK1|Integrin-linked protein kinase|ILK2|p59ILK |59 kDa serine/threonine-protein kinase|ILK-1|ILK-2
- Modification:phospho-Thr173
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:160-175/452
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Receptor-proximal protein kinase regulating integrin-mediated signal transduction. May act as a mediator of inside-out integrin signaling. Focal adhesion protein part of the complex ILK-PINCH. This complex is considered to be one of the convergence points of integrin- and growth factor-signaling pathway. Could be implicated in mediating cell architecture, adhesion to integrin substrates and anchorage-dependent growth in epithelial cells. Phosphorylates beta-1 and beta-3 integrin subunit on serine and threonine residues, but also AKT1 and GSK3B.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ILK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat