Order Entry
Denmark
ContactUsLinkComponent
Anti-NPM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-NPM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-4757R-A488
Supplier:  Bioss
Anti-NPM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-4757R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleophosmin
  • Antigen symbol:
    NPM
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    4869
  • Antigen synonyms:
    MGC104254|NO38|Nucleophosmin|Nucleolar protein NO38|numatrin|B23|Nucleolar phosphoprotein B23|NPM1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    1-50/259
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Involved in diverse cellular processes such as ribosome biogenesis, centrosome duplication, protein chaperoning, histone assembly, cell proliferation, and regulation of tumor suppressors p53/TP53 and ARF. Binds ribosome presumably to drive ribosome nuclear export. Associated with nucleolar ribonucleoprotein structures and bind single-stranded nucleic acids. Acts as a chaperonin for the core histones H3, H2B and H4. Stimulates APEX1 endonuclease activity on apurinic/apyrimidinic (AP) double-stranded DNA but inhibits APEX1 endonuclease activity on AP single-stranded RNA. May exert a control of APEX1 endonuclease activity within nucleoli devoted to repair AP on rDNA and the removal of oxidized rRNA molecules. In concert with BRCA2, regulates centrosome duplication. Regulates centriole duplication: phosphorylation by PLK2 is able to trigger centriole replication. Negatively regulates the activation of EIF2AK2/PKR and suppresses apoptosis through inhibition of EIF2AK2/PKR autophosphorylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat