Order Entry
Denmark
ContactUsLinkComponent
Anti-C3 Rabbit Polyclonal Antibody (Cy5®)
Anti-C3 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4876R-CY5
Supplier:  Bioss
Anti-C3 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4876R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement Component 3
  • Antigen symbol:
    C3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    718
  • Antigen synonyms:
    C3b|AHUS5|ARMD9|C3a|ASP|CPAMD1|HEL-S-62p
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The complement factor C3 consists of an alpha and a beta chain. C3 is a central factor in the complement cascade. It is central to the alternative pathway that leads to the C3 convertase C3bBb. The classical mannose binding lectin activation pathway leads to the C3 convertase C4b2a. These convertases cleave C3 resulting in C3a and C3b. Further degradation leads to the formation of the alpha chain products C3d, C3g and C3c. C3 is an acute phase protein that is produced by a wide range of tissues, including renal epithelial cells and hepatocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human