Order Entry
Denmark
ContactUsLinkComponent
Anti-BRLF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-BRLF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4500R-FITC
Supplier:  Bioss
Anti-BRLF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4500R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BRLF1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    180-230/605
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Immediate-early transcription factor that controls the initiation of viral lytic gene expression and lytic reactivation from latency. Triggers lytic replication, and initiates a cellular senescence program in epithelial cells. Upregulates human DCR3/TNFRSF6B by directly binding to its receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRLF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus