Order Entry
Denmark
ContactUsLinkComponent
Anti-MAP2K3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-MAP2K3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4123R-A350
Supplier:  Bioss
Anti-MAP2K3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4123R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-Activated Protein Kinase Kinase 3
  • Antigen symbol:
    MAP2K3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5606
  • Antigen synonyms:
    dual specificity mitogen-activated protein kinase kinase 3|SAPKK-2|MEK 3|MAP kinase kinase 3|MAPKK3|SAPK kinase 2|SAPKK2|PRKMK3|stress-activated protein kinase kinase 2|MKK3|MAPKK 3|MAPK/ERK kinase 3
  • Modification:
    phospho-Thr222
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Dual specificity kinase. Is activated by cytokines and environmental stress in vivo. Catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in the MAP kinase p38.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat