Anti-AAV5 VP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-10180R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Adeno-associated virus 5 capsid Protein VP1
- Antigen symbol:AAV5 VP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Capsid protein VP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
AAV5 capsid protein VP1
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AAV5 VP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus