Order Entry
Denmark
ContactUsLinkComponent
Anti-STK11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-STK11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-3948R-A555
Supplier:  Bioss
Anti-STK11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-3948R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serine/threonine Kinase 11
  • Antigen symbol:
    STK11
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6794
  • Antigen synonyms:
    PJS|Renal carcinoma antigen NY-REN-19|hLKB1|LKB1|STK11|Liver kinase B1|Serine/threonine-protein kinase STK11
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    30-90/433
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Tumor suppressor serine/threonine-protein kinase that controls the activity of AMP-activated protein kinase (AMPK) family members, thereby playing a role in various processes such as cell metabolism, cell polarity, apoptosis and DNA damage response. Acts by phosphorylating the T-loop of AMPK family proteins, thus promoting their activity: phosphorylates PRKAA1, PRKAA2, BRSK1, BRSK2, MARK1, MARK2, MARK3, MARK4, NUAK1, NUAK2, SIK1, SIK2, SIK3 and SNRK but not MELK. Also phosphorylates non-AMPK family proteins such as STRADA, PTEN and possibly p53/TP53. Acts as a key upstream regulator of AMPK by mediating phosphorylation and activation of AMPK catalytic subunits PRKAA1 and PRKAA2 and thereby regulates processes including: inhibition of signaling pathways that promote cell growth and proliferation when energy levels are low, glucose homeostasis in liver, activation of autophagy when cells undergo nutrient deprivation, and B-cell differentiation in the germinal center in response to DNA damage. Also acts as a regulator of cellular polarity by remodeling the actin cytoskeleton. Required for cortical neuron polarization by mediating phosphorylation and activation of BRSK1 and BRSK2, leading to axon initiation and specification. Involved in DNA damage response: interacts with p53/TP53 and recruited to the CDKN1A/WAF1 promoter to participate in transcription activation. Able to phosphorylate p53/TP53; the relevance of such result in vivo is however unclear and phosphorylation may be indirect and mediated by downstream STK11/LKB1 kinase NUAK1. Also acts as a mediator of p53/TP53-dependent apoptosis via interaction with p53/TP53: translocates to the mitochondrion during apoptosis and regulates p53/TP53-dependent apoptosis pathways. In vein endothelial cells, inhibits PI3K/Akt signaling activity and thus induces apoptosis in response to the oxidant peroxynitrite (in vitro). Regulates UV radiation-induced DNA damage response mediated by CDKN1A.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STK11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat