Anti-ZBTB8A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-4057R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger And BTB Domain Containing 8A
- Antigen symbol:ZBTB8A
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:653121
- Antigen synonyms:ZBTB8|ZNF916A|BOZF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB8A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat