- Antibody type:Primary
- Antigen name:UDP-Galactose-4-Epimerase
- Antigen symbol:GALE
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:2582
- Antigen synonyms:SDR1E1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
GALE is a 348 amino acid protein that functions as the third enzyme in the Leloir pathway of galactose metabolism. A member of the sugar epimerase family, GALE exists as a homodimer, binds FAD as a cofactor and catalyzes the epimerization of UDP-N-acetylglucosamine to UDP-N-acetylgalactosamine and UDP-glucose to UDP-galactose. The gene encoding GALE maps to human chromosome 1p36.11 and mutations in this gene lead to the development of complex disorder known as epimerase-deficiency galactosemia (EDG) or galactosemia type 3, which is characterized by mental retardation, liver damage, cataracts and deafness.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GALE/Galactowaldenase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat