Specifications
- Antibody type:Primary
- Antigen name:Galactose mutarotase
- Antigen symbol:GALM
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:130589
- Antigen synonyms:Galactose mutarotase|BLOCK25 protein|GALM|Aldose 1 epimerase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Galactose Mutarotase is a member of the aldose epimerase family and is involved in hexose metabolism. Through its catalytic activity, Galactose Mutarotase converts beta-aldose to alpha-aldose on several sugars, including D-glucose, L-arabinose and D-xylose. Found in the cytoplasm of most cells, Galactose Mutarotase plays a key role in galactose metabolism by catalyzing the conversion of beta-D-galactose to alpha-D-galactose. The enzyme contains two residues, Glu 304 and His 170, that are critical for catalysis, as well as His 96 and Asp 243, which are important for proper substrate recognition by the active site. No known diseases have been associated with mutations in the Galactose Mutarotase gene, although inhibition of Galactose Mutarotase activity could potentially be associated with a build-up of unmetabolized sugars during metabolism.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GALM/Galactose mutarotase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat