Order Entry
Denmark
ContactUsLinkComponent
 
Anti-SCN2B Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-12122R-CY7
 :  Bioss
Anti-SCN2B Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-12122R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Sodium Channel Voltage-gated Type II Beta
  • Antigen symbol:
    SCN2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    SCNB2|ATFB14
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Voltage-gated sodium channels are selective ion channels that regulate the permeability of sodium ions in excitable cells. During the propagation of an action potential, sodium channels allow an influx of sodium ions, which rapidly depolarizes the cell. Na+ CP type II beta(sodium channel, voltage-gated, type II, beta), also known as SCN2B, is a 215 amino acid single-pass type I membrane protein that plays a critical role in the expression and assembly of the heterotrimeric complex of the sodium channel and interacts with Tenascin-R to influence the clustering and regulation of sodium channels at nodes of Ranvier. Expressed specifically in brain, Na+ CP type II beta contains one Ig-like C2-type (immunoglobulin-like) domain and is encoded by a gene that maps to human chromosome 11q23.3 and mouse chromosome 9 A5.2.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCN2B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat