Anti-CDH3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-1159R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Cadherin 3
- Antigen symbol:CDH3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1001
- Antigen synonyms:Cdhp|AI385538|HJMD|CDHP|PCAD|Pcad|P-Cadherin|Cadp
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDH3
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: