Specifications
- Antibody type:Primary
- Antigen name:G Protein-coupled Receptor 143
- Antigen symbol:GPR143
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:OA1|NYS6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
G protein-coupled receptors (GPRs or GPCRs), are members of the largest protein family and play a role in many different stimulus-response pathways. G-protein coupled receptors mediate extracellular signals into intracellular signals (G-protein activation). They respond to a great variety of signaling molecules, including hormones, neurotransmitters and other proteins and peptides. GPR143, also designated ocular albinism type 1 protein (OA1), is detected exclusively in pigment cells. OA1, which is a multi-pass membrane protein, is a melanosomal protein expressed primarily in pigment cells. Defects in the gene encoding for OA1 cause ocular albinism, an X-linked disorder mainly characterized by retinal hypopigmentation and visual impairment.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OA1/GPR143
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat