Specifications
- Antibody type:Primary
- Antigen name:Dab, reelin signal transducer, homolog 1 (Drosophila)
- Antigen symbol:DAB1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:C630028C02Rik|scr|AI956902|scrambler|scm|yot
- Modification:phospho-Tyr198
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Dab1, a homolog of the Drosophila Disabled protein, is an adaptor protein involved in neural development. This cytoplasmic protein is tyrosine-phosphorylated during rapid expansion of the developing nervous system, and it is thought to interact with other proteins via a domain similar to the PTB domains of the Shc family. Dab1 has been shown to interact with the SH2 domains of Src, Fyn and Abl (1). Mutations in Dab1 result in widespread abnormalities in the brain, similar to those seen in Reelin mutants (2). Reelin is a secreted protein thought to play a role in directing migrating neurons (3). Evidence suggests that Dab1 functions downstream of Reelin in a signaling pathway involved in positioning cells in the developing brain (4). Dab2 (also designated DOC-2) is a mitogen-responsive phosphoprotein that binds the SH3 domain of Grb2, and it is thought to be a negative regulator of growth (5).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DAB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat