Order Entry
Denmark
ContactUsLinkComponent
Anti-B3GALNT1 Rabbit Polyclonal Antibody (Cy7®)
Anti-B3GALNT1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-10438R-CY7
Supplier:  Bioss
Anti-B3GALNT1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-10438R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
  • Antigen symbol:
    B3GALNT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8706
  • Antigen synonyms:
    GLOB|Gb4Cer|P|P1|galT3|B3GALT3|GLCT3|beta3Gal-T3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). The encoded protein of this gene does not use N-acetylglucosamine as an acceptor sugar at all. Multiple transcript variants that are alternatively spliced in the 5' UTR have been described; they all encode the same protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: B3GALNT1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat