Specifications
- Antibody type:Primary
- Antigen name:Insulin Receptor Substrate 2
- Antigen symbol:IRS2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Cross adsorption:No
- Form:liquid
- Gene ID:8660
- Antigen synonyms:IRS2|IRS-2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:540-590/1338
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The family of insulin receptor substrates (IRSs) has been reported to play important roles for signal transduction of various hormones. Four members of the IRS family have been described. Each IRS is believed to have different functions; however, the distinct physiological roles of each IRS are unclear. Summary: This gene encodes the insulin receptor substrate 2, a cytoplasmic signaling molecule that mediates effects of insulin, insulin-like growth factor 1, and other cytokines by acting as a molecular adaptor between diverse receptor tyrosine kinases and downstream effectors. The product of this gene is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation, as well as by an interleukin 4 receptor-associated kinase in response to IL4 treatment.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IRS2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat