Redox standards
Supplier: REAGECON
Jump to:
About this item
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
8 Options Available Below
Product Details & Documents
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- PkAvailabilityPrice
- REGERS124Standard redox solution, 124 mV at 25 °C
- REGERS200Standard redox solution, 200 mV at 25 °C
- REGERS250Standard redox solution, 250 mV at 25 °C
- REGERS300Standard redox solution, 300 mV at 25 °C
- REGERS358Standard redox solution, 358 mV at 25 °C
- REGERS465Standard redox solution, 465 mV at 25 °C
- REGERS600Standard redox solution, 600 mV at 25 °C
- REGERS650Standard redox solution, 650 mV at 25 °C
This product has been blocked by your organization. Please contact your purchasing department for more information