In-line MicroFilters, Upchurch Scientific®
These in-line MicroFilters effectively protect columns from particles originating in the mobile phase or sample, or from pump seal and sample injection valve wear.
- 100% biocompatible PEEK polymer options available
- Total void volume of 240 nl
- Pressure rated to 4000 psi (276 bar)
Frequently Bought Together