Pressure relief valves, Upchurch Scientific®
Pressure relief valves are ideal for preventing system over-pressurisation. These products protect system components by diverting fluid flow automatically when inline pressure exceeds the set limit.
: Supplied with flangeless fittings. The U-456 assembly is additionally supplied with wrenches for tightening.
Frequently Bought Together