Safety helmets, G3501
Supplier: 3M
Certificates
About this item
The G3501 electrically insulating 3M™ industrial safety helmet is designed to offer high levels of protection for the toughest of applications. The lightweight and durable helmet is also the platform for a range of hearing, eye and face protection products that are designed to withstand the metal working environments without compromising compatibility, comfort or ease of use.
- Compatible with a wide range of safety products and metal-working accessories to provide the complete solution for your application
- Available in a range of 6 colours for branding and identification purposes
Size: 530 to 620 mm, weight: 392 g.
Certifications: PPE Directive (89/686/EEC); EN 397:2012; EN 50365:2002 for live-working in low voltage environments, approved for use at temperatures ranging from –30 to +150 °C.
Frequently Bought Together