

Acetic acid ≥100%, LiChropur™ for LC-MS, Supelco®
5.33001.0050
: Merck
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Assay (acidimetric) :≥ 99.8 %
- Color :≤ 5 Hazen
- Al (Aluminium) :≤ 2.0 ppb
- Ca (Calcium) :≤ 20.0 ppb
- Cu (Copper) :≤ 1.0 ppb
- Fe (Iron) :≤ 20.0 ppb
- K (Potassium) :≤ 2.0 ppb
- Mg (Magnesium) :≤ 2.0 ppb
- Na (Sodium) :≤ 5.0 ppb
- NH4 (Ammonium) :≤ 10 ppb
- Residue on ignition (as sulfate) :≤ 2 ppb
- Intensity of background mass peak based on reserpine (ESI positive) :≤ 2 ppb
- Intensity of background mass peak based on reserpine (APCI negative) :≤ 20 ppb
:
:60.05 g/mol
:116…118 °C
:17 °C
:1.05 g/cm³
:39 °C
:Ambient
:200-580-7
:2789
:8 (3),II
:M-Clarity™ quality level = MQ100
:GHS02
:GHS05
:64-19-7
:H₃CCOOH
:MFCD00036152