Artemisinin ≥98%
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: Qinghaosu, Arteannuin, (3R,5aS,6R,8aS,9R,12S,12aR)-3,6,9-Trimethyloctahydro-12H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10(3H)-one
CAS Number:63968-64-9
:
Formula:C₁₅H₂₂O₅
MDL Number:MFCD00081057
Molecular Weight:282.34 g/mol
:GHS02
:GHS09
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together