Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
Formic acid 98 - 100% 98-100%, LiChropur™ for LC-MS, Supelco®
  5.33002.0050
 :  Merck
 :  
 
 
 
Formic acid 98 - 100% 98-100%, LiChropur™ for LC-MS, Supelco®
  5.33002.0050
 :  Merck
 :  

 

  • Pk:
    50 mL
  • Seq:
    2370

 

 

 :
 :46.03 g/mol
 :Ambient
 :200-579-1
 :1779
 :8 (3),II
 :GHS02
 :GHS06
 :GHS05
 :64-18-6
 :HCO₂H
 :MFCD00003297