Anti-CCL22 Rabbit Polyclonal Antibody
BWRLBS7709
- Antibody type:Primary
- Antigen symbol:CCL22
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human CCL22.
CCL22 pAb detects endogenous levels of CCL22 protein.
Type: Primary
Antigen: CCL22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human