Anti-RBPJ Rabbit Polyclonal Antibody
BWRLBS7661
- Antibody type:Primary
- Antigen name:Recombination signal binding protein for immunoglobulin kappa J
- Antigen symbol:RBPJ
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human RBPJ.
RBPJ pAb detects endogenous levels of RBPJ protein.
Type: Primary
Antigen: RBPJ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat