Anti-Cleaved-Cathepsin A Rabbit Polyclonal Antibody [clone: R326]
BWRLBS7038
- Antibody type:Primary
- Antigen symbol:Cleaved-Cathepsin A
- Clonality:Polyclonal
- Clone:R326
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 291-340 of Human Cleaved-Cathepsin A.
Cleaved-Cathepsin A (R326) pAb detects endogenous levels of Lysosomal protective protein 32 kDa chain.
Type: Primary
Antigen: Cleaved-Cathepsin A
Clonality: Polyclonal
Clone: R326
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat