Anti-CASP6 p18 Rabbit Polyclonal Antibody [clone: D162]
Catalog # BWRLBS7006
Supplier: Bioworld Technology
Specifications
- Antistof type:Primary
- Antigen symbol:CASP6 p18
- Clonality:Polyclonal
- Clone:D162
- Vært:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 141-190 of Human Caspase-6.
Caspase-6 p18 (D162) pAb detects endogenous levels of the large subunit of active caspase-6 resulting from cleavage at Asp162 in mouse and rat (18 kDa). This antibody does not recognize other cleaved caspases.
Type: Primary
Antigen: CASP6 p18
Clonality: Polyclonal
Clone: D162
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat