Anti-CA1 Rabbit Polyclonal Antibody
BWRLBS6959
- Antibody type:Primary
- Antigen symbol:CA1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human CA1.
CA1 pAb detects endogenous levels of CA1 protein.
Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat