Anti-PSMD7 Rabbit Polyclonal Antibody
BWRLBS7553
- Antistof type:Primary
- Antigen symbol:PSMD7
- Clonality:Polyclonal
- Vært:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Recombinant full length Human PSMD7.
PSMD7 pAb detects endogenous levels of PSMD7 protein.
Type: Primary
Antigen: PSMD7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat