Anti-VSV-G-Tag Mouse Monoclonal Antibody
Catalog # BWRLMB2021
Supplier: Bioworld Technology
Specifications
- Antistof type:Primary
- Antigen symbol:VSV-G-Tag
- Clonality:Monoclonal
- Vært:Mouse
- ImmunFluorescens:Yes
- Reactivity:Multiple
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Synthetic peptide VSV-G-Tag.
VSV-G-tag mouse monoclonal antibody detects endogenous levels of internal, C-terminal, and N-terminal VSV-G- tagged proteins.
Type: Primary
Antigen: VSV-G-Tag
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: All