Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Raf-B Mouse Monoclonal Antibody
Catalog # BWRLMB0117
Anti-Raf-B Mouse Monoclonal Antibody
Catalog # BWRLMB0117

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Raf-B
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human Raf-B.

Raf-B mAb detects endogenous levels of Raf-B protein.

Type: Primary
Antigen: Raf-B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse