Anti-p-VE-Cadherin Rabbit Polyclonal Antibody [clone: Y731]
BWRLBS4242
- Antibody type:Primary
- Antigen symbol:p-VE-Cadherin
- Clonality:Polyclonal
- Clone:Y731
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human VE-Cadherin around the phosphorylation site of Tyrosine 731.
p-VE-Cadherin (Y731) pAb detects endogenous levels of VE-Cadherin protein only when phosphorylated at Tyr731
Type: Primary
Antigen: p-VE-Cadherin
Clonality: Polyclonal
Clone: Y731
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat