Order Entry
Denmark
ContactUsLinkComponent
 
Anti-p-Rb Rabbit Polyclonal Antibody [clone: S795]
Catalog # BWRLBS4165
Anti-p-Rb Rabbit Polyclonal Antibody [clone: S795]
Catalog # BWRLBS4165

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Rb
  • Clonality:
    Polyclonal
  • Clone:
    S795
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Rb around the phosphorylation site of Serine 795.

p-Rb (S795) pAb detects endogenous levels of Rb protein only when phosphorylated at Ser795

Type: Primary
Antigen: p-Rb
Clonality: Polyclonal
Clone: S795
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat