Anti-Cytoglobin Rabbit Polyclonal Antibody
BWRLBS60396
- Antibody type:Primary
- Antigen symbol:Cytoglobin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
A synthetic peptide corresponding to residues in Human Cytoglobin.
Cytoglobin pAb detects endogenous levels of Cytoglobin protein.
Type: Primary
Antigen: Cytoglobin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat