Anti-Filaggrin Rabbit Polyclonal Antibody
BWRLBS60392
- Antibody type:Primary
- Antigen symbol:Filaggrin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
A synthetic peptide corresponding to residues in Human Filaggrin.
Filaggrin pAb detects endogenous levels of Filaggrin protein.
Type: Primary
Antigen: Filaggrin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat