Order Entry
Denmark
ContactUsLinkComponent
 
Anti-GHRHR Rabbit Polyclonal Antibody
  BWRLBS5732
Anti-GHRHR Rabbit Polyclonal Antibody
  BWRLBS5732

 

  • Antibody type:
    Primary
  • Antigen symbol:
    GHRHR
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 378-423 of Human GHRHR.

GHRHR pAb detects endogenous levels of GHRHR protein.

Type: Primary
Antigen: GHRHR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat