Anti-LZTR1 Rabbit Polyclonal Antibody
Catalog # BWRLBS60279
Supplier: Bioworld Technology
Specifications
- Antistof type:Primary
- Antigen symbol:LZTR1
- Clonality:Polyclonal
- Vært:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human LZTR1.
LZTR1 pAb detects endogenous levels of LZTR1 protein.
Type: Primary
Antigen: LZTR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat