Anti-FKHR/FOXO1A Rabbit Polyclonal Antibody [clone: S262]
Catalog # BWRLBS5518
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:FKHR/FOXO1A
- Clonality:Polyclonal
- Clone:S262
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 250-300 of Human FKHR.
FKHR/FOXO1A (S262) pAb detects endogenous levels of FKHR/FOXO1A protein.
Type: Primary
Antigen: FKHR/FOXO1A
Clonality: Polyclonal
Clone: S262
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat