Anti-CIB4 Rabbit Polyclonal Antibody
Catalog # BWRLBS60122
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CIB4
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human CIB4.
CIB4 pAb detects endogenous levels of CIB4 protein.
Type: Primary
Antigen: CIB4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat