Anti-p-NF2/Merlin Rabbit Polyclonal Antibody [clone: S518]
Catalog # BWRLBS4800
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-NF2/Merlin
- Clonality:Polyclonal
- Clone:S518
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Merlin around the phosphorylation site of Serine 518.
p-NF2/Merlin (S518) pAb detects endogenous levels of NF2/Merlin protein only when phosphorylated at Ser518
Type: Primary
Antigen: p-NF2/Merlin
Clonality: Polyclonal
Clone: S518
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat