Anti-PRMT5 Rabbit Polyclonal Antibody
Catalog # BWRLBS6835
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PRMT5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human PRMT5.
PRMT5 pAb detects endogenous levels of PRMT5 protein.
Type: Primary
Antigen: PRMT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human